siRNA Details
| Gene Symbol: Suclg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019879.3 | NP_063932.2 | 7 | 962 | ||
| GenBank mRNA | AF144101.2 | - | 7 | 795 | ||
| AK002754.1 | - | 7 | 797 | |||
| AK005080.1 | - | 7 | 811 | |||
| AK088888.1 | - | 7 | 819 | |||
| BC011087.1 | - | 7 | 783 | 4012310 | ||

Suclg1
Genomic Map