siRNA Details
| Gene Symbol: Clptm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019649.2 | NP_062623.2 | 7, 8 | 819 | ||
| GenBank mRNA | AK148080.1 | - | 7, 8 | 801 | ||
| AK152410.1 | - | 7, 8 | 809 | |||
| AK152809.1 | - | 7, 8 | 809 | |||
| AK165742.1 | - | 6, 7 | 1513 | |||
| AK171901.1 | - | 7, 8 | 810 | |||
| AK181139.1 | - | 2, 3 | 291 | |||
| BC022172.1 | - | 7, 8 | 810 | 4193279 | ||
| D67067.1 | - | 7, 8 | 806 | |||

Clptm1
Genomic Map