siRNA Details
| Gene Symbol: Snd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019776.2 | NP_062750.2 | 20, 21 | 2514 | ||
| GenBank mRNA | AB021491.1 | - | 20, 21 | 2471 | ||
| AK088028.1 | - | 20, 21 | 2473 | |||
| AK133837.1 | - | 20, 21 | 2520 | |||
| AK159837.1 | - | 20, 21 | 2479 | |||
| AK161591.1 | - | 20, 21 | 2468 | |||
| AK162432.1 | - | 20, 21 | 2455 | |||
| AK181132.1 | - | 3, 4 | 248 | |||
| BC007126.1 | - | 20, 21 | 2461 | 3592548 | ||

Snd1
Genomic Map