siRNA Details
| Gene Symbol: Pias1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019663.3 | NP_062637.2 | 6 | 795 | ||
| GenBank mRNA | AF077950.1 | - | 6 | 787 | ||
| AK013753.1 | - | 2 | 130 | |||
| AK075708.1 | - | 6 | 796 | |||
| AK153871.1 | - | 6 | 800 | |||
| BC051417.1 | - | 6 | 792 | 6515027 | ||

Pias1
Genomic Map