siRNA Details
| Gene Symbol: Foxo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019740.2 | NP_062714.1 | 3 | 1677 | ||
| GenBank mRNA | AF114259.1 | - | 3 | 1677 | ||
| AK047413.1 | - | 3 | 1635 | |||
| AK139629.1 | - | 3 | 1665 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxo3| Gene Symbol: Foxo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019740.2 | NP_062714.1 | 3 | 1677 | ||
| GenBank mRNA | AF114259.1 | - | 3 | 1677 | ||
| AK047413.1 | - | 3 | 1635 | |||
| AK139629.1 | - | 3 | 1665 | |||