siRNA Details
| Gene Symbol: Nxt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110159.1 | NP_001103629.1 | 3 | 547 | ||
| NM_019761.6 | NP_062735.4 | 2 | 438 | |||
| GenBank mRNA | AF156958.2 | - | 2 | 379 | ||
| AK003254.1 | - | 2 | 414 | |||
| AK168314.1 | - | 2 | 398 | |||
| BC023672.1 | - | 2 | 396 | 5039139 | ||

Nxt1
Genomic Map