siRNA Details
| Gene Symbol: Ikbke | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019777.3 | NP_062751.2 | 21 | 2406 | ||
| GenBank mRNA | AB016589.1 | - | 19 | 2094 | ||
| AK088580.1 | - | 21 | 2408 | |||
| AK149911.1 | - | 21 | 2395 | |||
| AK153046.1 | - | 21 | 2407 | |||
| AK180824.1 | - | 2 | 42 | |||
| AK220324.1 | - | 4 | 4778 | |||
| BC037446.1 | - | 20 | 2078 | 3980145 | ||
| CT010206.1 | - | 19 | 2060 | |||
| CT010313.1 | - | 19 | 1988 | |||

Ikbke
Genomic Map