siRNA Details
| Gene Symbol: Ruvbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019685.2 | NP_062659.1 | 4 | 593 | ||
| GenBank mRNA | AF100694.1 | - | 4 | 632 | ||
| AK012170.1 | - | 4 | 541 | |||
| AK132783.1 | - | 4 | 575 | |||
| AK146258.1 | - | 4 | 585 | |||
| AK146374.1 | - | 4 | 586 | |||
| AK156078.1 | - | 4 | 563 | |||
| BC004718.1 | - | 4 | 559 | 3582426 | ||

Ruvbl1
Genomic Map