siRNA Details
| Gene Symbol: Rapgef4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204165.1 | NP_001191094.1 | 14 | 1430 | ||
| NM_001204166.1 | NP_001191095.1 | 11 | 1322 | |||
| NM_019688.2 | NP_062662.1 | 13 | 1376 | |||
| GenBank mRNA | AB021132.1 | - | 14 | 1405 | ||
| AB037668.1 | - | 5 | 765 | |||
| AB445011.1 | - | 11 | 1314 | |||
| AF115480.1 | - | 13 | 1304 | |||
| AK220281.1 | - | 14 | 1430 | |||
| BC139822.1 | - | 14 | 1362 | 40141811 | ||

Rapgef4
Genomic Map