siRNA Details
| Gene Symbol: Rnf138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB025010.1 | - | 5 | 738 | ||
| AB025011.1 | - | 5 | 764 | |||
| AK004238.1 | - | 4 | 540 | |||
| AK010486.1 | - | 4 | 529 | |||
| AK136138.1 | - | 4 | 541 | |||
| BC003712.1 | - | 4 | 504 | 3593899 | ||

Rnf138
Genomic Map