siRNA Details
| Gene Symbol: Mpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164733.1 | NP_001158205.1 | 13, 14 | 1759 | ||
| NM_001164734.1 | NP_001158206.1 | 13, 14 | 1769 | |||
| NM_019939.2 | NP_064323.2 | 12, 13 | 1727 | |||
| GenBank mRNA | AF161181.1 | - | 12, 13 | 1591 | ||
| AF199009.1 | - | 12, 13 | 1723 | |||
| AF199010.1 | - | 13, 14 | 1765 | |||
| AK077754.1 | - | 3, 4 | 345 | |||
| AK144434.1 | - | 13, 14 | 1770 | |||
| BC138666.1 | - | 12, 13 | 1585 | 8861687 | ||
| BC145363.1 | - | 13, 14 | 1627 | 9053921 | ||
| BC145364.1 | - | 13, 14 | 1617 | 9053922 | ||

Mpp6
Genomic Map