siRNA Details
| Gene Symbol: Zfp235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019941.2 | NP_064325.2 | 4 | 1121 | ||
| GenBank mRNA | AB221682.1 | - | 2 | 753 | ||
| AF167321.1 | - | 4 | 1085 | |||
| AK147833.1 | - | 5 | 1300 | |||
| BC099965.1 | - | 4 | 1042 | 30084081 | ||

Zfp235
Genomic Map