siRNA Details
| Gene Symbol: Cnpy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019953.1 | NP_064337.1 | 4 | 650 | ||
| GenBank mRNA | AF186115.1 | - | 4 | 650 | ||
| AK007914.1 | - | 4 | 411 | |||
| AK013014.1 | - | 4 | 521 | |||
| AK013568.1 | - | 4 | 392 | |||
| AK019927.1 | - | 4 | 522 | |||
| AK150953.1 | - | 4 | 594 | |||
| BC008261.1 | - | 4 | 422 | 2650612 | ||

Cnpy2
Genomic Map