siRNA Details
| Gene Symbol: Ylpm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178363.3 | NP_848140.3 | 9 | 5253 | ||
| GenBank mRNA | AB033168.1 | - | 9 | 2926 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ylpm1| Gene Symbol: Ylpm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178363.3 | NP_848140.3 | 9 | 5253 | ||
| GenBank mRNA | AB033168.1 | - | 9 | 2926 | ||