siRNA Details
| Gene Symbol: Habp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019986.3 | NP_064370.2 | 6 | 1012 | ||
| GenBank mRNA | AF227684.1 | - | 6 | 955 | ||
| AK016800.1 | - | 6 | 1055 | |||
| AK144206.1 | - | 6 | 996 | |||
| AK146525.1 | - | 6 | 985 | |||
| BC082806.1 | - | 6 | 1014 | |||
| BC119460.2 | - | 5 | 508 | |||
| BC148304.2 | - | 5 | 516 | |||

Habp4
Genomic Map