siRNA Details
| Gene Symbol: Ick | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163780.1 | NP_001157252.1 | 12 | 2053 | ||
| NM_019987.2 | NP_064371.2 | 12 | 1981 | |||
| GenBank mRNA | AF225918.1 | - | 12 | 1821 | ||
| AK052178.1 | - | 5 | 950 | |||
| AK087484.1 | - | 12 | 1905 | |||
| AK133537.1 | - | 12 | 1920 | |||
| AK154185.1 | - | 12 | 2054 | |||
| AK173068.1 | - | 7 | 1210 | |||
| AK197692.1 | - | 1 | 31 | |||
| BC028863.1 | - | 11 | 1738 | 4224269 | ||

Ick
Genomic Map