siRNA Details
Gene Symbol: Ube2d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019912.2 | NP_064296.1 | 5 | 723 | ||
GenBank mRNA | AK005355.1 | - | 6 | 675 | ||
AK136197.1 | - | 5 | 661 | |||
AK139623.1 | - | 5 | 689 | |||
AK146659.1 | - | 3 | 855 | |||
AK147568.1 | - | 4 | 6409 | |||
AK148089.1 | - | 5 | 545 | |||
AK160567.1 | - | 5 | 696 | |||
AK162093.1 | - | 5 | 528 | |||
AK172088.1 | - | 5 | 550 | |||
AK216283.1 | - | 1 | 103 | |||
AK217918.1 | - | 11 | ||||
BC003923.1 | - | 5 | 704 | 3485259 | ||
U62483.1 | - | 5 | 679 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031001, NM_001037292, BC078808, U56407, U13176.
Panther Classification:
Gene Ontology Categories:
