siRNA Details
| Gene Symbol: Mgst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019946.4 | NP_064330.2 | 4 | 334 | ||
| GenBank mRNA | AF159050.1 | - | 3 | 233 | ||
| AK002800.1 | - | 4 | 335 | |||
| AK005122.1 | - | 4 | 335 | |||
| BC009155.1 | - | 3 | 273 | 2655738 | ||

Mgst1
Genomic Map