siRNA Details
| Gene Symbol: Trim17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031172.2 | NP_112449.1 | 7 | 1298 | ||
| GenBank mRNA | AF220135.1 | - | 1 | 58 | ||
| AK077212.1 | - | 7 | 1281 | |||
| BC055112.1 | - | 7 | 1298 | 6707359 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim17| Gene Symbol: Trim17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031172.2 | NP_112449.1 | 7 | 1298 | ||
| GenBank mRNA | AF220135.1 | - | 1 | 58 | ||
| AK077212.1 | - | 7 | 1281 | |||
| BC055112.1 | - | 7 | 1298 | 6707359 | ||