siRNA Details
| Gene Symbol: Prl2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019991.1 | NP_064375.1 | 5 | 623 | ||
| GenBank mRNA | AB032428.1 | - | 5 | 644 | ||
| AF226610.1 | - | 5 | 579 | |||
| AF234636.1 | - | 5 | 623 | |||
| AF272368.1 | - | 5 | 575 | |||
| AK005424.1 | - | 5 | 600 | |||
| BC099458.1 | - | 5 | 619 | 30790018 | ||

Prl2a1
Genomic Map