siRNA Details
| Gene Symbol: Klk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019928.1 | NP_064312.1 | 4 | 423 | ||
| GenBank mRNA | AF019979.1 | - | 4 | 423 | ||
| AK087426.1 | - | 4 | 437 | |||
| BC100716.1 | - | 3 | 395 | 40045068 | ||
| BC100717.1 | - | 3 | 395 | 40045067 | ||
| BC100718.1 | - | 3 | 395 | 40045065 | ||
| BC100719.1 | - | 3 | 395 | 40045063 | ||

Klk4
Genomic Map