siRNA Details
| Gene Symbol: Ankrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020033.1 | NP_064417.1 | 8 | 790 | ||
| GenBank mRNA | AB076256.1 | - | 8 | 802 | ||
| AJ011118.1 | - | 8 | 790 | |||
| AJ245514.1 | - | 8 | 790 | |||
| AK132585.1 | - | 8 | 800 | |||
| BC138760.1 | - | 8 | 828 | 8861782 | ||
| BC138761.1 | - | 8 | 828 | 8861783 | ||

Ankrd2
Genomic Map