siRNA Details
| Gene Symbol: Slc15a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053079.2 | NP_444309.2 | 17 | 1318 | ||
| GenBank mRNA | AF205540.1 | - | 17 | 1317 | ||
| AK085471.1 | - | 15 | 2425 | |||
| AK136428.1 | - | 17 | 1319 | |||
| BC116248.1 | - | 17 | 1308 | 40093541 | ||
| BC116249.1 | - | 17 | 1308 | 40093542 | ||

Slc15a1
Genomic Map