siRNA Details
| Gene Symbol: Clec7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020008.2 | NP_064392.2 | 4 | 564 | ||
| GenBank mRNA | AA240643.1 | - | 3 | 407 | ||
| AF262985.1 | - | 4 | 560 | |||
| AY534909.1 | - | 4 | 472 | |||
| BC027742.1 | - | 4 | 563 | 5036647 | ||

Clec7a
Genomic Map