siRNA Details
| Gene Symbol: Dnajb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019964.1 | NP_064348.1 | 1 | 505 | ||
| GenBank mRNA | AB028856.1 | - | 1 | 505 | ||
| AK006026.1 | - | 1 | 489 | |||
| BC049591.1 | - | 1 | 483 | 6773959 | ||
| BC061112.1 | - | 1 | 488 | 6772873 | ||

Dnajb8
Genomic Map