siRNA Details
| Gene Symbol: Crtap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019922.2 | NP_064306.2 | 4 | 1022 | ||
| GenBank mRNA | AJ006469.1 | - | 4 | 980 | ||
| AK007624.1 | - | 4 | 743 | |||
| AK017797.1 | - | 4 | 985 | |||
| AK047506.1 | - | 4 | 978 | |||
| AK203540.1 | - | 2 | 162 | |||
| BC049890.1 | - | 4 | 959 | 6306169 | ||

Crtap
Genomic Map