siRNA Details
| Gene Symbol: Crtap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019922.2 | NP_064306.2 | 1 | 550 | ||
| GenBank mRNA | AJ006469.1 | - | 1 | 508 | ||
| AK017797.1 | - | 1 | 513 | |||
| AK047506.1 | - | 1 | 506 | |||
| BC049890.1 | - | 1 | 487 | 6306169 | ||

Crtap
Genomic Map