siRNA Details
Gene Symbol: Gpr132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019925.4 | NP_064309.1 | 3 | 1048 | ||
GenBank mRNA | AF083442.1 | - | 3 | 1084 | ||
AK089866.1 | - | 3 | 1025 | |||
AK153765.1 | - | 3 | 1049 | |||
AK154509.1 | - | 3 | 1070 | |||
AK157600.1 | - | 3 | 1075 | |||
BC120522.1 | - | 2 | 975 | 40129445 | ||
BC120524.1 | - | 2 | 975 | 40129447 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
