siRNA Details
Gene Symbol: Hist1h1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020034.2 | NP_064418.1 | 1 | 687 | ||
GenBank mRNA | AK020117.1 | - | 1 | 688 | ||
BC114579.1 | - | 1 | 615 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hist1h1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020034.2 | NP_064418.1 | 1 | 687 | ||
GenBank mRNA | AK020117.1 | - | 1 | 688 | ||
BC114579.1 | - | 1 | 615 |