siRNA Details
| Gene Symbol: Rabgef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199059.1 | NP_001185988.1 | 9 | 1307 | ||
| NM_019983.2 | NP_064367.1 | 8 | 1452 | |||
| GenBank mRNA | AF093590.1 | - | 9 | 1283 | ||
| AK044568.1 | - | 8 | 1472 | |||
| AK146734.1 | - | 9 | 1257 | |||
| BC018229.1 | - | 8 | 1452 | 4507477 | ||

Rabgef1
Genomic Map