siRNA Details
| Gene Symbol: Litaf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019980.2 | NP_064364.1 | 2 | 166 | ||
| GenBank mRNA | AF171100.1 | - | 2 | 134 | ||
| AF220207.1 | - | 1 | 88 | |||
| AF230522.1 | - | 1 | 116 | |||
| AK018578.1 | - | 2 | 149 | |||
| AK076162.1 | - | 2 | 168 | |||
| AK078708.1 | - | 2 | 166 | |||
| AK151053.1 | - | 2 | 166 | |||
| AK159533.1 | - | 2 | 134 | |||
| BC018559.1 | - | 2 | 146 | 2812674 | ||

Litaf
Genomic Map