siRNA Details
| Gene Symbol: Cript | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019936.3 | NP_064320.1 | 2 | 163 | ||
| GenBank mRNA | AF151637.1 | - | 2 | 128 | ||
| AK004828.1 | - | 2 | 156 | |||
| AK008523.1 | - | 2 | 148 | |||
| AK029409.1 | - | 2 | 151 | |||
| BC023013.1 | - | 2 | 137 | 5369938 | ||
| BY123761.1 | - | 2 | 164 | |||

Cript
Genomic Map