siRNA Details
| Gene Symbol: Krt71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019956.1 | NP_064340.1 | 6, 7 | 1160 | ||
| GenBank mRNA | AB033744.1 | - | 6, 7 | 1160 | ||
| AB100413.1 | - | 6, 7 | 1160 | |||
| AK003610.1 | - | 6, 7 | 1171 | |||
| AK003704.1 | - | 6, 7 | 1172 | |||
| AY033497.1 | - | 6, 7 | 1160 | |||
| BC125346.1 | - | 6, 7 | 1155 | 40129861 | ||
| BC125348.1 | - | 6, 7 | 1155 | 40129863 | ||

Krt71
Genomic Map