siRNA Details
| Gene Symbol: Mocs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020042.2 | NP_064426.2 | 11 | 1519 | ||
| NM_028464.1 | NP_082740.1 | 11 | 1534 | |||
| GenBank mRNA | AF214016.1 | - | 5 | 709 | ||
| AK157231.1 | - | 11 | 1475 | |||
| BC027444.1 | - | 3 | 445 | |||
| BC034066.1 | - | 7 | 883 | |||
| BC049914.1 | - | 9 | 1162 | |||
| BC147411.1 | - | 2 | 363 | |||
| BC147412.1 | - | 2 | 348 | |||
| BC172067.1 | - | 2 | 348 | |||

Mocs1
Genomic Map