siRNA Details
| Gene Symbol: Rec8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020002.3 | NP_064386.2 | 6, 7 | 713 | ||
| GenBank mRNA | AF262055.1 | - | 6, 7 | 643 | ||
| AK077167.1 | - | 6, 7 | 648 | |||
| AK146832.1 | - | 6, 7 | 699 | |||
| BC052155.1 | - | 6, 7 | 709 | 6335959 | ||

Rec8
Genomic Map