siRNA Details
| Gene Symbol: Igdcc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290315.1 | NP_001277244.1 | 13 | 2387 | ||
| NM_020043.3 | NP_064427.2 | 13 | 2384 | |||
| GenBank mRNA | AB052620.1 | - | 13 | 2384 | ||
| AB052621.1 | - | 13 | 2387 | |||
| AF176694.1 | - | 13 | 2335 | |||
| AK122535.1 | - | 2463 | ||||
| BU701299.1 | - | 3 | 377 | |||

Igdcc4
Genomic Map