siRNA Details
| Gene Symbol: Nfu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170591.1 | NP_001164062.1 | 3, 4 | 370 | ||
| NM_020045.3 | NP_064429.2 | 3, 4 | 370 | |||
| GenBank mRNA | AJ132616.1 | - | 3, 4 | 301 | ||
| AK002244.1 | - | 3, 4 | 316 | |||
| AK003730.1 | - | 3, 4 | 318 | |||
| AK145558.1 | - | 3, 4 | 371 | |||
| AY335195.1 | - | 3, 4 | 277 | |||
| BC018355.1 | - | 3, 4 | 291 | 3584656 | ||

Nfu1
Genomic Map