siRNA Details
| Gene Symbol: Tacstd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020047.3 | NP_064431.2 | 1 | 780 | ||
| GenBank mRNA | AK076410.1 | - | 1 | 755 | ||
| AK077370.1 | - | 1 | 755 | |||
| AK085753.1 | - | 1 | 755 | |||
| BC117719.1 | - | 1 | 744 | 40088161 | ||
| BC117720.1 | - | 1 | 744 | 40088163 | ||

Tacstd2
Genomic Map