siRNA Details
| Gene Symbol: Clec1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204239.1 | NP_001191168.1 | 2 | 436 | ||
| NM_001204253.1 | NP_001191182.1 | 2 | 436 | |||
| NM_019985.3 | NP_064369.1 | 3 | 532 | |||
| GenBank mRNA | AF201457.1 | - | 3 | 243 | ||
| AK007925.1 | - | 3 | 512 | |||
| BC064054.1 | - | 3 | 532 | 574821 | ||
| EF070191.1 | - | 2 | 126 | |||
| EF070192.1 | - | 2 | 123 | |||

Clec1b
Genomic Map