siRNA Details
| Gene Symbol: Scube2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020052.2 | NP_064436.2 | 3, 4 | 462 | ||
| GenBank mRNA | AK142950.1 | - | 3, 4 | 428 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scube2| Gene Symbol: Scube2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020052.2 | NP_064436.2 | 3, 4 | 462 | ||
| GenBank mRNA | AK142950.1 | - | 3, 4 | 428 | ||