siRNA Details
| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019995.3 | NP_064379.1 | 5, 6 | 351 | ||
| GenBank mRNA | AF139179.1 | - | 5, 6 | 355 | ||
| AK212629.1 | - | 4, 5 | 208 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Supt20| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019995.3 | NP_064379.1 | 5, 6 | 351 | ||
| GenBank mRNA | AF139179.1 | - | 5, 6 | 355 | ||
| AK212629.1 | - | 4, 5 | 208 | |||