siRNA Details
| Gene Symbol: Ube2l6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019949.2 | NP_064333.2 | 3 | 374 | ||
| GenBank mRNA | AF159230.1 | - | 3 | 360 | ||
| AK010942.1 | - | 3 | 353 | |||
| AK013452.1 | - | 3 | 364 | |||
| AK152010.1 | - | 3 | 354 | |||
| AK152066.1 | - | 3 | 326 | |||
| AK152113.1 | - | 3 | 375 | |||
| AK168695.1 | - | 3 | 312 | |||
| AK171629.1 | - | 3 | 323 | |||
| BC008238.1 | - | 3 | 342 | 2651702 | ||
| CT010270.1 | - | 3 | 293 | |||

Ube2l6
Genomic Map