siRNA Details
| Gene Symbol: Ube2l6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019949.2 | NP_064333.2 | 4 | 415 | ||
| GenBank mRNA | AF159230.1 | - | 4 | 401 | ||
| AK010942.1 | - | 4 | 394 | |||
| AK013452.1 | - | 4 | 405 | |||
| AK152010.1 | - | 4 | 395 | |||
| AK152066.1 | - | 4 | 367 | |||
| AK152113.1 | - | 4 | 416 | |||
| AK168695.1 | - | 4 | 353 | |||
| AK171629.1 | - | 4 | 364 | |||
| BC008238.1 | - | 4 | 383 | 2651702 | ||
| CT010270.1 | - | 4 | 334 | |||

Ube2l6
Genomic Map