siRNA Details
| Gene Symbol: Dkk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020265.4 | NP_064661.2 | 3, 4 | 1268 | ||
| GenBank mRNA | AJ243963.2 | - | 3, 4 | 668 | ||
| AK028225.1 | - | 3, 4 | 553 | |||
| AK031749.1 | - | 3, 4 | 1269 | |||
| BC096448.1 | - | 3, 4 | 1259 | 6389874 | ||

Dkk2
Genomic Map