siRNA Details
| Gene Symbol: Ccl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020279.3 | NP_064675.1 | 2 | 164 | ||
| GenBank mRNA | AF220238.1 | - | 2 | 110 | ||
| BC040510.1 | - | 2 | 176 | |||
| BC055864.1 | - | 2 | 167 | 4240191 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccl28| Gene Symbol: Ccl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020279.3 | NP_064675.1 | 2 | 164 | ||
| GenBank mRNA | AF220238.1 | - | 2 | 110 | ||
| BC040510.1 | - | 2 | 176 | |||
| BC055864.1 | - | 2 | 167 | 4240191 | ||