siRNA Details
| Gene Symbol: Lgi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020278.2 | NP_064674.1 | 8 | 1140 | ||
| GenBank mRNA | AK031996.1 | - | 8 | 1050 | ||
| AK045443.1 | - | 8 | 1245 | |||
| BC066090.1 | - | 8 | 1140 | 6839091 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lgi1| Gene Symbol: Lgi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020278.2 | NP_064674.1 | 8 | 1140 | ||
| GenBank mRNA | AK031996.1 | - | 8 | 1050 | ||
| AK045443.1 | - | 8 | 1245 | |||
| BC066090.1 | - | 8 | 1140 | 6839091 | ||