siRNA Details
| Gene Symbol: Aldh1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053080.3 | NP_444310.3 | 13 | 1559 | ||
| GenBank mRNA | AF152359.1 | - | 13 | 1514 | ||
| AF246711.1 | - | 13 | 1507 | |||
| AF253409.1 | - | 13 | 1546 | |||
| AF280404.1 | - | 13 | 1511 | |||
| AK147006.1 | - | 13 | 1560 | |||
| BC038646.1 | - | 4 | 380 | |||
| BC048941.1 | - | 5 | 472 | |||
| BC058277.1 | - | 13 | 1503 | 6515355 | ||

Aldh1a3
Genomic Map