siRNA Details
| Gene Symbol: Olfr690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020290.2 | NP_064686.2 | 1 | 876 | ||
| GenBank mRNA | BC137757.1 | - | 1 | 876 | 8860776 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr690| Gene Symbol: Olfr690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020290.2 | NP_064686.2 | 1 | 876 | ||
| GenBank mRNA | BC137757.1 | - | 1 | 876 | 8860776 | |