siRNA Details
| Gene Symbol: Nsmf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039386.1 | NP_001034475.1 | 14, 15 | 1653 | ||
| NM_001039387.1 | NP_001034476.1 | 13, 14 | 1647 | |||
| NM_001177655.1 | NP_001171126.1 | 12, 13 | 1557 | |||
| NM_020276.3 | NP_064672.2 | 13, 14 | 1584 | |||
| GenBank mRNA | AF266508.2 | - | 12, 13 | 1550 | ||
| AJ278902.1 | - | 14, 15 | 1431 | |||
| AK004986.2 | - | 13, 14 | 1644 | |||
| AK019560.1 | - | 14, 15 | 1620 | |||
| AK053402.1 | - | 12, 13 | 1504 | |||
| AK158803.1 | - | 14, 15 | 1383 | |||
| BC006642.1 | - | 13, 14 | 1605 | 3483536 | ||

Nsmf
Genomic Map