siRNA Details
| Gene Symbol: Htr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020274.4 | NP_064670.1 | 6, 7 | 882 | ||
| GenBank mRNA | AF155045.1 | - | 6, 7 | 714 | ||
| AK085213.1 | - | 6, 7 | 883 | |||
| BC046990.1 | - | 6, 7 | 860 | 6309747 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Htr3b| Gene Symbol: Htr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020274.4 | NP_064670.1 | 6, 7 | 882 | ||
| GenBank mRNA | AF155045.1 | - | 6, 7 | 714 | ||
| AK085213.1 | - | 6, 7 | 883 | |||
| BC046990.1 | - | 6, 7 | 860 | 6309747 | ||